accessibility menu, dialog, popup

UserName

H3CH3CNNNHOHOH3CH3CCH3HOCH3H3CCH3

CAS RN 758679-97-9

IUPAC Name: 1-(3,5-di-tert-butyl-4-hydroxyphenyl)-2-{2-[(3-hydroxypropyl)amino]-5,6-dimethyl-1H-1,3-benzodiazol-1-yl}ethan-1-one
Synonyms: 1-(3,5-di-tert-butyl-4-hydroxyphenyl)-2-{2-[(3-hydroxypropyl)amino]-5,6-dimethyl-1,3-benzodiazol-1-yl}ethanone
Molecular Weight (g/mol): 465.64
Molecular Formula: C28H39N3O3
InChi Key: CYCGGKILBWERDJ-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C2N(CC(=O)C3=CC(=C(O)C(=C3)C(C)(C)C)C(C)(C)C)C(NCCCO)=NC2=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results