accessibility menu, dialog, popup

UserName

CAS RN 76189-56-5

PP(?)

CAS RN 76189-56-5

IUPAC Name: [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane
Synonyms: binap
Molecular Weight (g/mol): 622.69
Molecular Formula: C44H32P2
InChi Key: MUALRAIOVNYAIW-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (5)
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)