accessibility menu, dialog, popup

UserName

CAS RN 76287-49-5

H3CBr

CAS RN 76287-49-5

IUPAC Name: 1-bromo-4-heptylbenzene
Synonyms: 1-bromo-4-heptylbenzene
Molecular Weight (g/mol): 255.2
Molecular Formula: C13H19Br
InChi Key: YHKMTIJHJWYYAG-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results