accessibility menu, dialog, popup

UserName

CHF

CAS RN 766-49-4

IUPAC Name: 1-ethynyl-2-fluorobenzene
Synonyms: 1-ethynyl-2-fluorobenzene
Molecular Weight (g/mol): 120.13
Molecular Formula: C8H5F
InChi Key: YFPQIXUNBPQKQR-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results