accessibility menu, dialog, popup

UserName

H3CNOO

CAS RN 76620-00-3

IUPAC Name: 1-(4-ethylphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-ethylphenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: FHVHFKZQDVQILM-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)N1C(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results