missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Ethylphenyl)maleimide, 98+%

Product Code. 11354434
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354434 10 g 10g
11344434 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354434 Supplier Thermo Scientific Alfa Aesar Supplier No. L01058.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 76620-00-3 | C12H11NO2 | 201.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76620-00-3
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.225
MDL Number MFCD00014538
InChI Key FHVHFKZQDVQILM-UHFFFAOYSA-N
Synonym n-4-ethylphenyl maleimide, 1-4-ethylphenyl-1h-pyrrole-2,5-dione, 1-4-ethylphenyl pyrrole-2,5-dione, 1-4-ethylphenyl maleimide, 1-4-ethylphenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-4-ethylphenyl azoline-2,5-dione, 1-4-ethylphenyl-1h-pyrrole-2,5-dione #, 1h-pyrrole-2,5-dione, 1-4-ethylphenyl
PubChem CID 522624
IUPAC Name 1-(4-ethylphenyl)pyrrole-2,5-dione
SMILES CCC1=CC=C(C=C1)N2C(=O)C=CC2=O

Specifications

Melting Point 64°C to 67°C
Quantity 10 g
Beilstein 7022079
Formula Weight 201.22
Percent Purity ≥98%
Chemical Name or Material N-(4-Ethylphenyl)maleimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.