accessibility menu, dialog, popup

UserName

CAS RN 768-32-1

IUPAC Name: trimethyl(phenyl)silane
Synonyms: silane, trimethyl phenyl
Molecular Weight (g/mol): 150.3
Molecular Formula: C9H14Si
InChi Key: KXFSUVJPEQYUGN-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results