missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylsilane, 98%

Product Code. 11397875
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397875 5 g 5g
11307885 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397875 Supplier Thermo Scientific Alfa Aesar Supplier No. L02876.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 768-32-1 | C9H14Si | 150.30 g/mol

It is employed as intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical and organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-32-1
Molecular Formula C9H14Si
Molecular Weight (g/mol) 150.30
MDL Number MFCD00008271
InChI Key KXFSUVJPEQYUGN-UHFFFAOYSA-N
Synonym phenyltrimethylsilane, trimethyl phenyl silane, silane, trimethylphenyl, trimethylsilyl benzene, unii-px1sg1q5yp, benzene, trimethylsilyl, silane, trimethyl phenyl, px1sg1q5yp, phenyl trimethylsilane, trimethyl-phenylsilane
PubChem CID 69849
IUPAC Name trimethyl(phenyl)silane
SMILES C[Si](C)(C)C1=CC=CC=C1

Specifications

Boiling Point 169°C to 170°C
Quantity 5 g
UN Number UN1993
Beilstein 1855012
Flash Point 40°C (104°F)
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 150.3
Percent Purity 98%
Refractive Index 1.49
Density 0.871
Chemical Name or Material Phenyltrimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.