accessibility menu, dialog, popup

UserName

CAS RN 768-33-2

CH3SiH3CCl

CAS RN 768-33-2

IUPAC Name: chlorodimethylphenylsilane
Synonyms: chlorodimethylphenylsilane
Molecular Weight (g/mol): 170.71
Molecular Formula: C8H11ClSi
InChi Key: KWYZNESIGBQHJK-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)