missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylphenylsilane, 97+%

Product Code. 11477537
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477537 100 g 100g
11457537 5 g 5g
11467537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477537 Supplier Thermo Scientific Alfa Aesar Supplier No. A15638.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 768-33-2 | C8H11ClSi | 170.71 g/mol

Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

Solubility
Miscible with tetrahydrofuran, ether and hexane.

Notes
Incompatible with strong oxidizing agents, acids, bases, and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-33-2
Molecular Formula C8H11ClSi
Molecular Weight (g/mol) 170.71
MDL Number MFCD00000499
InChI Key KWYZNESIGBQHJK-UHFFFAOYSA-N
Synonym chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
PubChem CID 13029
IUPAC Name chloro-dimethyl-phenylsilane
SMILES C[Si](C)(Cl)C1=CC=CC=C1

Specifications

Boiling Point 192°C to 193°C
Quantity 100 g
UN Number UN2987
Beilstein 606292
Flash Point 72°C (161°F)
Solubility Information Miscible with tetrahydrofuran,ether and hexane.
Formula Weight 170.72
Percent Purity ≥97%
Refractive Index 1.509
Sensitivity Moisture sensitive
Density 1.032
Chemical Name or Material Chlorodimethylphenylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.