accessibility menu, dialog, popup

UserName

HNNNNH2OHSOOHO

CAS RN 769951-32-8

IUPAC Name: 7H-pyrrolo[2,3-d]pyrimidin-4-amine; sulfuric acid
Synonyms: 7-deazaadenine; sulfuric acid
Molecular Weight (g/mol): 232.21
Molecular Formula: C6H8N4O4S
InChi Key: ZVFLCRGXDSPVKK-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=C2C=CNC2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results