missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-7-deazapurine hydrogen sulfate, 97%, Thermo Scientific™

Product Code. 15451879
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451879 250 mg 250mg
15431879 1 g 1g
15441879 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451879 Supplier Thermo Scientific Alfa Aesar Supplier No. H64602.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769951-32-8
Molecular Formula C6H8N4O4S
Molecular Weight (g/mol) 232.214
MDL Number MFCD09787504
InChI Key ZVFLCRGXDSPVKK-UHFFFAOYSA-N
Synonym 4-amino-7h-pyrrolo 2,3-d pyrimidine hydrogen sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulphate, 4-amino-7h-pyrrolo 2,3-d pyrimidine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine; sulfuric acid, 7-deazaadenine; sulfuric acid, 1h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfuric acid, 4-amino-7h-pyrrolo 2,3-d pyrimidinehydrogensulfate, ksc376q6n
PubChem CID 11470294
IUPAC Name 7H-pyrrolo[2,3-d]pyrimidin-4-amine;sulfuric acid
SMILES C1=CNC2=C1C(=NC=N2)N.OS(=O)(=O)O

Specifications

Color Brown
Quantity 250 mg
Formula Weight 232.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Amino-7-deazapurine hydrogen sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.