accessibility menu, dialog, popup

UserName

TiIIII

CAS RN 7720-83-4

IUPAC Name: titanium(4+) tetraiodide
Synonyms: titanium tetraiodide
Molecular Weight (g/mol): 555.49
Molecular Formula: I4Ti
InChi Key: NLLZTRMHNHVXJJ-UHFFFAOYSA-J
SMILES: [Ti+4].[I-].[I-].[I-].[I-]
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results