missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) iodide, ultra dry, 99.99% (metals basis)

Product Code. 11318587
Change view
Click to view available options
Quantity:
2 g
Packaging:
Ampule
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11318587 2 g Ampule 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11318587 Supplier Thermo Scientific Alfa Aesar Supplier No. 035724.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7720-83-4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 150°C
CAS 7720-83-4
Physical Form Powder
Quantity 2 g
MDL Number MFCD00043077
UN Number UN3260
Formula Weight 555.52
Percent Purity 99.99%
Odor Acrid
Packaging Ampule
Sensitivity Moisture sensitive
Density 4.30 g/mL
Assay (metals basis)
Chemical Name or Material Titanium(IV) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.