accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNHOH

CAS RN 77279-24-4

IUPAC Name: 4-[(tert-butoxy)carbonyl]-1-(2-hydroxyethyl)piperazin-1-ium
Synonyms: 4-(tert-butoxycarbonyl)-1-(2-hydroxyethyl)piperazin-1-ium
Molecular Weight (g/mol): 231.32
Molecular Formula: C11H23N2O3
InChi Key: VRXIOAYUQIITBU-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)N1CC[NH+](CCO)CC1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results