missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-hydroxyethyl)piperazine, 97%

Product Code. 15422408
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422408 25 g 25g
15402408 1 g 1g
15412408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422408 Supplier Thermo Scientific Alfa Aesar Supplier No. H55991.14
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 77279-24-4 | C11H22N2O3 | 230.308 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77279-24-4
Molecular Formula C11H22N2O3
Molecular Weight (g/mol) 230.308
MDL Number MFCD00728947
InChI Key VRXIOAYUQIITBU-UHFFFAOYSA-N
Synonym tert-butyl 4-2-hydroxyethyl piperazine-1-carboxylate, 1-boc-4-2-hydroxyethyl piperazine, 1-tert-butyloxy carbonyl-4-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-hydroxyethyl-piperazine, 4-2-hydroxy-ethyl-piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-2-hydroxyethyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-2-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl-1-piperazinecarboxylate, 4-2-hydroxyethyl piperazine-1-carboxylic acid tert-butyl ester
PubChem CID 735736
IUPAC Name tert-butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCO

Specifications

Melting Point 38°C to 42°C
Color Brown to White
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN2811
Formula Weight 230.31
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(2-hydroxyethyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.