accessibility menu, dialog, popup

UserName

CAS RN 77284-32-3

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoate
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoate
Molecular Weight (g/mol): 352.41
Molecular Formula: C21H22NO4
InChi Key: VCFCFPNRQDANPN-LJQANCHMSA-M
SMILES: CCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)