accessibility menu, dialog, popup

UserName

NH2FFF

CAS RN 774-18-5

IUPAC Name: 2-[2-(trifluoromethyl)phenyl]ethan-1-amine
Synonyms: 2-[2-(trifluoromethyl)phenyl]ethanamine
Molecular Weight (g/mol): 189.18
Molecular Formula: C9H10F3N
InChi Key: LIERORLYMWHXDL-UHFFFAOYSA-N
SMILES: NCCC1=CC=CC=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results