accessibility menu, dialog, popup

UserName

CAS RN 77451-51-5

CH3SOONNNNOO

CAS RN 77451-51-5

IUPAC Name: 1-(4-methylbenzenesulfonyl)-3-nitro-1H-1,2,4-triazole
Synonyms: 1-(4-methylbenzenesulfonyl)-3-nitro-1,2,4-triazole
Molecular Weight (g/mol): 268.25
Molecular Formula: C9H8N4O4S
InChi Key: ZQMJAWSQRGYFBM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results