missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole, 98%, Thermo Scientific™

Product Code. 11976861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976861 5 g 5g
11966861 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976861 Supplier Thermo Scientific Acros Supplier No. 223250050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 77451-51-5
Molecular Formula C9H8N4O4S
Molecular Weight (g/mol) 268.25
MDL Number MFCD00009756
InChI Key ZQMJAWSQRGYFBM-UHFFFAOYSA-N
Synonym 3-nitro-1-tosyl-1h-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-methylphenyl sulfonyl-3-nitro-1,2,4-triazole, 1-4-methylbenzenesulfonyl-3-nitro-1,2,4-triazole, tsnt, 3-nitro-1-tosyl-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazol, 3-nitro-1-p-tolylsulfonyl-1,2,4-triazole, 3-nitro-1-p-toluenesulfonyl-1,2,4-triazole
PubChem CID 688072
IUPAC Name 1-(4-methylphenyl)sulfonyl-3-nitro-1,2,4-triazole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Assay Percent Range 98%
Formula Weight 268.25
Quantity 5 g
Infrared Spectrum Authentic
Physical Form Powder or Crystalline Powder
Chemical Name or Material 1-(p-Toluenesulfonyl)-3-nitro-1, 2, 4-triazole
compliance-icons
Product Identifier
  • 1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.