accessibility menu, dialog, popup

UserName

BNNHNNOHOH

CAS RN 775351-30-9

IUPAC Name: [3-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
Synonyms: 3-(2H-1,2,3,4-tetrazol-5-yl)phenylboronic acid
Molecular Weight (g/mol): 189.97
Molecular Formula: C7H7BN4O2
InChi Key: XMJKBDSKRITXBW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results