missing translation for 'onlineSavingsMsg'
Learn More

3-(Tetrazol-5-yl)phenylboronic acid, 96%, Thermo Scientific Chemicals

Product Code. 11916761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916761 1 g 1g
11926761 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916761 Supplier Thermo Scientific Acros Supplier No. 445560010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775351-30-9
Molecular Formula C7H7BN4O2
Molecular Weight (g/mol) 189.97
MDL Number MFCD11044436
InChI Key XMJKBDSKRITXBW-UHFFFAOYSA-N
Synonym 3-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, boronic acid,b-3-2h-tetrazol-5-yl phenyl, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, acmc-209pal
PubChem CID 46737996
IUPAC Name [3-(2H-tetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC=C1)C1=NNN=N1

Specifications

Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.97
Percent Purity 96%
Chemical Name or Material 3-(Tetrazol-5-yl)phenylboronic acid
compliance-icons
Product Identifier
  • 3-(Tetrazol-5-yl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.