accessibility menu, dialog, popup

UserName

H2OCH3ONHOHOOHHOHO

CAS RN 7772-94-3

IUPAC Name: N-[2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide hydrate
Synonyms: N-acetylmannosamine
Molecular Weight (g/mol): 239.22
Molecular Formula: C8H17NO7
InChi Key: VVQPUTSNIMAJPT-UHFFFAOYNA-N
SMILES: O.CC(=O)NC1C(O)OC(CO)C(O)C1O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)