missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-mannosamine, 98%, Thermo Scientific Chemicals

Product Code. 10527970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10527970 5 g Glass bottle 5g
10351470 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10527970 Supplier Thermo Scientific Acros Supplier No. 427530050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N-Acetyl-D-mannosamine (ManNAc), CAS # 7772-94-3, is a monosaccharide sialic acid precursor.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • N-Acetyl-D-mannosamine (ManNAc) is a precursor used to synthesize N-acetylneuraminic acid, the most common physiologic sialic acid present in humans and other mammals
  • Sialic acids are terminal sugars on glycoconjugates that can modify biochemistry of glycoproteins and lipids, mediate cellular adhesion, and act as immune regulators
  • Defects in sialic acid biosynthesis are associated with a number of pathologies
  • Sialic acids are often found at increased expression levels on tumor cell surfaces

Applications

  • As a sialic acid precursor, N-Acetyl-D-mannosamine can be used in many glycoengineering applications, such as modifying immune cell activity, imaging glycans, or blocking pathogen binding
  • N-Acetyl-D-mannosamine hasbeen used in preclinical laboratory studies as a treatment for hereditary inclusion body myopathy, a progressive neuromuscular disease via increased sialylation of podocalyxin
  • N-Acetyl-D-mannosamine has been used in laboratory studies to induce IgG sialylation and prevent obesity-induced hypertension in mice following high-fat diet
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-94-3
Molecular Formula C8H17NO7
Molecular Weight (g/mol) 239.22
MDL Number MFCD00149493,MFCD00046758,MFCD00149493,MFCD00069808
InChI Key VVQPUTSNIMAJPT-UHFFFAOYNA-N
Synonym n-acetylmannosamine
PubChem CID 11908605
IUPAC Name N-[(2R,3S,4S,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES O.CC(=O)NC1C(O)OC(CO)C(O)C1O

Spécification

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 221.21
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-D-mannosamine
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.