accessibility menu, dialog, popup

UserName

OHSeOHO

CAS RN 7783-00-8

IUPAC Name: selenous acid
Synonyms: selenious acid monohydrated selenium dioxide selenious acid preparation H2SeO3 selenious acid (H2seO3) acid, selenious selenium (as selenious acid) preparation selenige saeure dihydroxidooxidoselenium selenium (as selenious acid)
Molecular Weight (g/mol): 128.98
Molecular Formula: H2O3Se
InChi Key: MCAHWIHFGHIESP-UHFFFAOYSA-N
SMILES: O[Se](O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results