CAS RN 7783-11-1
IUPAC Name:
diammonium hydrate sulfite
Synonyms:
diammonium hydrate sulfite
Molecular Weight (g/mol):
134.15
Molecular Formula:
H10N2O4S
InChi Key:
ASPXBYAQZVXSNS-UHFFFAOYSA-N
SMILES:
[NH4+].[NH4+].O.[O-]S([O-])=O
1
–
2
of
2
results
1
Ammonium sulfite monohydrate, 92% min
CAS: 7783-11-1 Molecular Formula: H10N2O4S Molecular Weight (g/mol): 134.15 MDL Number: MFCD00066094 InChI Key: ASPXBYAQZVXSNS-UHFFFAOYSA-N PubChem CID: 21701053 IUPAC Name: diazanium;sulfite;hydrate SMILES: [NH4+].[NH4+].O.[O-]S(=O)[O-]