missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfite monohydrate, Honeywell Fluka™

Product Code. 15615040 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15615040 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615040 Supplier Honeywell-Fluka Supplier No. 358983500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥92%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-11-1
Quantity 500 g
Molecular Formula H10N2O4S
Linear Formula (NH4)2SO3 · H2O
MDL Number MFCD00150959
InChI Key ASPXBYAQZVXSNS-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].O.[O-]S(=O)[O-]
IUPAC Name diazanium;sulfite;hydrate
Molecular Weight (g/mol) 134.15
PubChem CID 21701053
Formula Weight 134.16g/mol
Concentration or Composition (by Analyte or Components) 92%
Density 1.41g/mL at 25°C
Chemical Name or Material Ammonium sulfite monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.