accessibility menu, dialog, popup

UserName

CAS RN 7783-97-3

AgOIOO

CAS RN 7783-97-3

IUPAC Name: silver(1+) iodate
Synonyms: silver iodate
Molecular Weight (g/mol): 282.77
Molecular Formula: AgIO3
InChi Key: YSVXTGDPTJIEIX-UHFFFAOYSA-M
SMILES: [Ag+].[O-][I](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results