missing translation for 'onlineSavingsMsg'
Learn More

Silver iodate, 98%, Thermo Scientific Chemicals

Product Code. 11399237
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11399237 Supplier Thermo Scientific Alfa Aesar Supplier No. 040109.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-97-3
Molecular Formula AgIO3
Molecular Weight (g/mol) 282.77
MDL Number MFCD00014150
InChI Key YSVXTGDPTJIEIX-UHFFFAOYSA-M
Synonym silver iodate, unii-5p7ox4vt1f, silver 1+ ion iodate, 5p7ox4vt1f, silberjodat, silver 1+ iodate, acmc-20ajqa, iodic acid hio3 , silver 1+ salt, ag.io3, silver iodate 25g
PubChem CID 165642
IUPAC Name silver;iodate
SMILES [O-]I(=O)=O.[Ag+]

Specifications

Melting Point >200°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Practically insoluble in water. Soluble in aqueous ammonia
Formula Weight 282.77
Odor Odorless
Sensitivity Light sensitive
Appearance White Powder
Density 5.53 g/cm3 at 20°C
Chemical Name or Material Silver iodate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.