accessibility menu, dialog, popup

UserName

CrONOOONOOONOO

CAS RN 7789-02-8

IUPAC Name: chromium(3+) trinitrate
Synonyms: chromium nitrate
Molecular Weight (g/mol): 238.01
Molecular Formula: CrN3O9
InChi Key: PHFQLYPOURZARY-UHFFFAOYSA-N
SMILES: [Cr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (15)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (8)
  • (1)
Physical Form 
  • (6)
  • (5)
  • (1)
  • (1)
Grade 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (8)
  • (15)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (8)
  • (1)
  • (6)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)