accessibility menu, dialog, popup

UserName

BrPBrBrBrBr

CAS RN 7789-69-7

IUPAC Name: pentabromo-λ⁵-phosphane
Synonyms: phosphorus pentabromide
Molecular Weight (g/mol): 430.49
Molecular Formula: Br5P
InChi Key: QRKVRHZNLKTPGF-UHFFFAOYSA-N
SMILES: BrP(Br)(Br)(Br)Br
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)