missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) bromide, 95%

Product Code. 11420667
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420667 500 g 500g
11400667 25 g 25g
11410667 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420667 Supplier Thermo Scientific Alfa Aesar Supplier No. B22481.36
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7789-69-7 | Br5P | 430.494 g/mol

Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-69-7
Molecular Formula Br5P
Molecular Weight (g/mol) 430.494
MDL Number MFCD00011437
InChI Key QRKVRHZNLKTPGF-UHFFFAOYSA-N
Synonym phosphorus pentabromide, phosphorane, pentabromo, unii-3d9wis0bqw, pentabromophosphorane, phosphorus v bromide, 3d9wis0bqw, pentabromo, phosphorpentabromid, phosphoruspentabromide, phosphorouspentabromide
PubChem CID 62678
IUPAC Name pentabromo-$l^{5}-phosphane
SMILES P(Br)(Br)(Br)(Br)Br

Specifications

Boiling Point 105°C to 107°C (decomposition)
Melting Point ∽105°C (decomposition)
Quantity 500 g
Odor Acrid
Sensitivity Moisture sensitive
UN Number UN2691
Beilstein 3903332
Merck Index 14,7350
Solubility Information Reacts with water.
Formula Weight 430.5
Percent Purity 95%
Chemical Name or Material Phosphorus(V) bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.