accessibility menu, dialog, popup

UserName

CH3POCH3CH3

CAS RN 78-50-2

IUPAC Name: 1-(dioctylphosphoryl)octane
Synonyms: trioctylphosphine oxide
Molecular Weight (g/mol): 386.65
Molecular Formula: C24H51OP
InChi Key: ZMBHCYHQLYEYDV-UHFFFAOYSA-N
SMILES: CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)