missing translation for 'onlineSavingsMsg'
Learn More

Trioctylphosphine Oxide, BAKER™, J.T. Baker™

Product Code. 15518844 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15518844 100 g Wide Mouth Amber Glass Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518844 Supplier Avantor J.T.Baker Supplier No. X00505

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Specifications

CAS 78-50-2
Flash Point 252.00°C
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C24H51OP
Quantity 100 g
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
IUPAC Name 1-dioctylphosphoryloctane
Molecular Weight (g/mol) 386.645
PubChem CID 65577
Formula Weight 386.65
Chemical Name or Material Trioctylphosphine Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.