CAS RN 780-31-4
IUPAC Name:
methyl 4-(trifluoromethoxy)benzoate
Synonyms:
methyl 4-(trifluoromethoxy)benzoate
Molecular Weight (g/mol):
220.15
Molecular Formula:
C9H7F3O3
InChi Key:
JFFLBWZTJAWGNP-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(OC(F)(F)F)C=C1
1
–
2
of
2
results
1
Methyle4-(trifluoromethoxy)benzoate, 97%, Thermo Scientific™
CAS: 780-31-4 Molecular Formula: C9H7F3O3 Molecular Weight (g/mol): 220.147 MDL Number: MFCD00041006 InChI Key: JFFLBWZTJAWGNP-UHFFFAOYSA-N PubChem CID: 737353 IUPAC Name: methyl 4-(trifluoromethoxy)benzoate SMILES: COC(=O)C1=CC=C(C=C1)OC(F)(F)F