missing translation for 'onlineSavingsMsg'
Learn More

Methyle4-(trifluoromethoxy)benzoate, 97%, Thermo Scientific™

Product Code. 10569972
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10569972 1 g 1g
10492494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10569972 Supplier Thermo Scientific Maybridge Supplier No. TL00423DA
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 780-31-4
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD00041006
InChI Key JFFLBWZTJAWGNP-UHFFFAOYSA-N
PubChem CID 737353
IUPAC Name methyl 4-(trifluoromethoxy)benzoate
SMILES COC(=O)C1=CC=C(C=C1)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4C(O)OCH3
Quantity 1 g
Synonym methyl 4-trifluoromethoxy benzoate, benzoic acid, 4-trifluoromethoxy-, methyl ester, acmc-209pd6, methyl4-trifluoromethoxy benzoate, 2-trifluoromethoxy-terephthalonitrile, 4-trifluoromethoxybenzoic acid methyl ester, p-trifluoromethoxy benzoic acid methyl ester, benzoic acid,4-trifluoromethoxy-, methyl ester
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material Methyle4-(trifluoromethoxy)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.