accessibility menu, dialog, popup

UserName

CAS RN 783-13-1

IUPAC Name: (2E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoate
Synonyms: (2E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoate
Molecular Weight (g/mol): 231.15
Molecular Formula: C10H6F3O3
InChi Key: RNYVTJANWYBGPW-ZZXKWVIFSA-M
SMILES: [O-]C(=O)\C=C\C1=CC=C(OC(F)(F)F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results