accessibility menu, dialog, popup

UserName

CH3NONNNNONNNCH3H3CH3CCH3

CAS RN 783331-24-8

IUPAC Name: 5-({[3-tert-butyl-7-(5-methyl-1,2-oxazol-3-yl)pyrazolo[1,5-d][1,2,4]triazin-2-yl]oxy}methyl)-1-methyl-1H-1,2,4-triazole
Synonyms: 5-({[3-tert-butyl-7-(5-methyl-1,2-oxazol-3-yl)pyrazolo[1,5-d][1,2,4]triazin-2-yl]oxy}methyl)-1-methyl-1,2,4-triazole
Molecular Weight (g/mol): 368.4
Molecular Formula: C17H20N8O2
InChi Key: QYSYOGCIDRANAR-UHFFFAOYSA-N
SMILES: CN1N=CN=C1COC1=NN2C(C=NN=C2C2=NOC(C)=C2)=C1C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results