accessibility menu, dialog, popup

UserName

CAS RN 78348-24-0

IUPAC Name: (2S)-2,3-dihydro-1H-indole-2-carboxylate
Synonyms: (2S)-2,3-dihydro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 162.17
Molecular Formula: C9H8NO2
InChi Key: QNRXNRGSOJZINA-QMMMGPOBSA-M
SMILES: [O-]C(=O)[C@@H]1CC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)