missing translation for 'onlineSavingsMsg'
Learn More

Indoline-2-carboxylic Acid, TRC

Product Code. 30405177
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405177 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405177 Supplier Toronto Research Chemicals Supplier No. TRCI6272051G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Indoline-2-carboxylic Acid
CAS 78348-24-0
Molecular Formula C9H9NO2
Quantity 1 g
Synonym (±)-2,3-Dihydroindole-2-carboxylic Acid, (±)-Indoline-2-carboxylic Acid, 2,3-Dihydro-1H-indole-2-carboxylic Acid, 2,3-Dihydroindole-2-carboxylic Acid, Indoline-2-carboxylic Acid
InChI Formula InChI=1S/C9H9NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h1-4,8,10H,5H2,(H,11,12)
SMILES OC(=O)C1Cc2ccccc2N1
IUPAC Name 2,3-dihydro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 163.17
Formula Weight 163.0633
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.