accessibility menu, dialog, popup

UserName

HOOO

CAS RN 78508-96-0

IUPAC Name: 5-(hydroxymethyl)-2,5-dihydrofuran-2-one
Synonyms: 5-(hydroxymethyl)-5H-furan-2-one
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: AWNLUIGMHSSXHB-UHFFFAOYNA-N
SMILES: OCC1OC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results