missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-5-Hydroxymethyl-2(5H)-furanone, 98%

Product Code. 11312717
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312717 250 mg 250mg
11302717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312717 Supplier Thermo Scientific Alfa Aesar Supplier No. L11919.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 78508-96-0 | C5H6O3 | 114.1 g/mol

(S)-(-)-5-Hydroxymethyl-2(5H)-furanone is used in the preparation of partially saturated heterocycles via a diastereoselective ring chain formation. It is also used in the synthesis of (1R,3S)-cis-chrysanthemic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone is used in the preparation of partially saturated heterocycles via a diastereoselective ring chain formation. It is also used in the synthesis of (1R,3S)-cis-chrysanthemic acid.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78508-96-0
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00075370
InChI Key AWNLUIGMHSSXHB-BYPYZUCNSA-N
Synonym s---5-hydroxymethyl-2 5h-furanone, s-5-hydroxymethyl furan-2 5h-one, 2s-2-hydroxymethyl-2h-furan-5-one, 2 5h-furanone, 5-hydroxymethyl-, 5s, 5s-5-hydroxymethyl-5h-furan-2-one, pubchem7063, s-5-hydroxy-2-penten-4-olide, 5-hydroxymethyl-2 5h-furanone #, 5beta-hydroxymethyl furan-2 5h-one
PubChem CID 10176122
IUPAC Name (2S)-2-(hydroxymethyl)-2H-furan-5-one
SMILES C1=CC(=O)OC1CO

Specifications

Melting Point 34°C to 43°C
Quantity 250 mg
Beilstein 3537455
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Optical Rotation −148° (c=1.4 in Water)
Formula Weight 114.1
Percent Purity 98%
Chemical Name or Material (S)-(-)-5-Hydroxymethyl-2(5H)-furanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.