accessibility menu, dialog, popup

UserName

CAS RN 789-25-3

Si

CAS RN 789-25-3

IUPAC Name: triphenylsilyl
Synonyms: triphenylsilyl
Molecular Weight (g/mol): 259.4
Molecular Formula: C18H15Si
InChi Key: BZLZKLMROPIZSR-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Si](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)