missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsilane, TRC

Product Code. 30454977
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30454977

Brand: Toronto Research Chemicals TRCT896008100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylsilane
CAS 789-25-3
Molecular Formula C18H16Si
Quantity 100 mg
InChI Formula InChI=1S/C18H16Si/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,19H
SMILES c1ccc(cc1)[SiH](c2ccccc2)c3ccccc3
IUPAC Name triphenylsilane
Molecular Weight (g/mol) 260.41
Formula Weight 260.1021
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.