accessibility menu, dialog, popup

UserName

HNSOOClOHO

CAS RN 78922-04-0

IUPAC Name: 3-(3-chlorobenzenesulfonamido)benzoic acid
Synonyms: 3-(3-chlorobenzenesulfonamido)benzoic acid
Molecular Weight (g/mol): 311.74
Molecular Formula: C13H10ClNO4S
InChi Key: APBOVLPLJFJSRI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(NS(=O)(=O)C2=CC(Cl)=CC=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)