missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IR120, H form, ion-exchange resin, Thermo Scientific™

Product Code. 10781604 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10781604

Brand: Thermo Scientific Acros 369720051

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Amberlite IR120, H form, ion-exchange resin, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 78922-04-0, Amber to Brown-Yellow, Molecular Formula: C13H10ClNO4S, Molecular Weight: 311.736

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78922-04-0
Molecular Formula C13H10ClNO4S
Molecular Weight (g/mol) 311.736
MDL Number MFCD00132707
InChI Key APBOVLPLJFJSRI-UHFFFAOYSA-N
Synonym 3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
PubChem CID 8190984
IUPAC Name 3-[(3-chlorophenyl)sulfonylamino]benzoic acid
SMILES C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient <1.8, harmonic mean size: 0.620 to 0.830mm,% <0.300mm: <2.0%,%>1.180mm: <3.0%
Packaging Plastic drum
Volume Total Capacity (H+):> 1.8 eq/L
Water Retention Capacity (H+): 53.0 to 58.0%
Color Amber to Brown/Yellow
Quantity 5 kg
Chemical Name or Material Amberlite™ IR120
compliance-icons
Product Identifier
  • Amberlite]TS IR120 H resin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.