accessibility menu, dialog, popup

UserName

H3C(R)CH3H3C(R)(R)H3C(R)CH3(S)CH3(S)HOH3CCH3(R)

CAS RN 79-63-0

IUPAC Name: (1R,3aR,5aR,7S,9aS,11aR)-3a,6,6,9a,11a-pentamethyl-1-[(2R)-6-methylhept-5-en-2-yl]-1H,2H,3H,3aH,4H,5H,5aH,6H,7H,8H,9H,9aH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms: lanosterol (3β)-lanosta-8,24-dien-3-ol
Molecular Weight (g/mol): 426.73
Molecular Formula: C30H50O
InChi Key: CAHGCLMLTWQZNJ-BQNIITSRSA-N
SMILES: C[C@H](CCC=C(C)C)[C@H]1CC[C@@]2(C)C3=C(CC[C@]12C)[C@@]1(C)CC[C@H](O)C(C)(C)[C@@H]1CC3
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results