missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lanosterol, 50%

Product Code. 17271156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17271156 25 g 25g
17261156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17271156 Supplier Thermo Scientific Chemicals Supplier No. 466720250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 79-63-0 | C30H50O, C30H50O | 426.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-63-0
Molecular Formula C30H50O

Specifications

Assay Percent Range 50%
Quantity 25 g
Melting Point 137.0°C
Formula Weight 426.73
Chemical Name or Material Lanosterol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.