accessibility menu, dialog, popup

UserName

BFFFFFFFFFFFFFFFFFFFFFFFFNa

CAS RN 79060-88-1

IUPAC Name: sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
Synonyms: sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
Molecular Weight (g/mol): 886.21
Molecular Formula: C32H12BF24Na
InChi Key: LTGMONZOZHXAHO-UHFFFAOYSA-N
SMILES: [Na+].FC(F)(F)C1=CC(=CC(=C1)[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results