accessibility menu, dialog, popup

UserName

OHSOHOOOSOO

CAS RN 8014-95-7

IUPAC Name: sulfonylideneoxidane; sulfuric acid
Synonyms: sulfur trioxide; sulfuric acid
Molecular Weight (g/mol): 178.13
Molecular Formula: H2O7S2
InChi Key: HIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILES: O=S(=O)=O.OS(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)